missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCIAD1 Mouse anti-Human, Mouse, Rat, Clone: 1C10C3, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66698-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody detects OCIAD1 with an apparent molecular weight of ∽35 kDa as has been demonstrated by several researches.
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. At least eight members of the Rab subfamily have been identified, each of which is found at a particular stage of a membrane transport pathway.Spécification
OCIAD1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5XIG4, Q9CRD0, Q9NX40 | |
Ociad1 | |
OCIAD1 Fusion Protein Ag9977 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG2a |
Western Blot | |
1C10C3 | |
Unconjugated | |
OCIAD1 | |
Asrij, FLJ20455, OCIA, OCIA domain containing 1, OCIAD1, TPA018 | |
Mouse | |
Protein A | |
RUO | |
289590, 54940, 68095 | |
-20°C | |
Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu