Learn More
LCORL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17468-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
May act as transcription activator that binds DNA elements with the sequence 5′-CCCTATCGATCGATCTCTACCT-3′. May play a role in spermatogenesis.Spécification
LCORL | |
Polyclonal | |
Unconjugated | |
LCORL | |
LCoR like protein, LCORL, MLR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209707, 254251, 498385 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3U285, Q8N3X6 | |
Lcorl | |
LCORL Fusion Protein Ag11377 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |