Learn More
DWK Life Sciences Kimble™ KIMAX™ Squibb Separatory Funnels with PTFE-Plug Stopcock
Do not require grease
Brand: DWK Life Sciences 29048F-500
This item is not returnable.
View return policy
Description
Do not require grease
Lower stems made of large-diameter tubing for liquid to drain out completely when stopcock is closed.
- Eliminate contamination
- Will not bind or freeze
Order Info
Stem lengths, 60mm.
Compliance
Designed from ASTM™ E 1096-86.
Specifications
500 mL | |
ASTM E1096, Type IV | |
60mm | |
4; PTFE | |
27 |
Borosilicate Glass | |
Eliminates contamination | |
Each | |
Glass | |
Squibb Separatory Funnel |