Learn More
IFIT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12604-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IFIT2 belongs to the IFIT family. It contains 6 TPR repeats. IFIT genes comprise a large family with three (Ifit1, Ifit2, and Ifit3) and four (IFIT1, IFIT2, IFIT3, and IFIT5) members in mice and humans, respectively. IFITproteins are produced in human body that are supposed to confer immunity against viral infection. These proteins are generally produced during viral infection, Interferon (IFN) treatment, and during pathogen recognition (Pathogen associated molecular pattern recognition) byimmune systemduring infections.Spécification
IFIT2 | |
Polyclonal | |
Unconjugated | |
IFIT2 | |
cig42, G10P2, GARG 39, IFI 54, IFI 54K, IFI54, IFIT 2, IFIT2, ISG 54 K, ISG 54K, ISG54 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15958, 3433 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09913, Q64112 | |
IFIT2 | |
IFIT2 Fusion Protein Ag3283 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |