missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.5M/l Solution, Scharlab

Product Code. 11322485
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in analytical chemistry as a titrant in volumetric analysis.

  • Molecular Formula: HCl
  • Molar Mass: 36.46g/mol
  • CAS: 7647-01-0
  • EIN ECS No.: 231-595-7
  • Molarity: 0.5 mol/l

This volumetric solution was checked by means of potentiometric methods using Scharlau's calcium carbonate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).

Specifications

CAS 7647-01-0
Composition 1mL = 0.018235g HCl
Concentration 0.5mol/L
Density 1.01g/cm3 
Traceability to NIST Traceable to NIST
Normality 0.5N
Quantity 1 L
Molecular Formula ClH
Physical Form Solution
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid, solution 0,5 mol/l (0,5 N)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.