Learn More
GBP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17746-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Guanylate-binding proteins, such as GBP4, are induced by interferon and hydrolyze GTP to both GDP and GMP.[supplied by OMIM, Dec 2008].Spécification
GBP4 | |
Polyclonal | |
Unconjugated | |
GBP4 | |
GBP 4, GBP4, GTP binding protein 4, guanylate binding protein 4, Mpa2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115361 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96PP9 | |
GBP4 | |
GBP4 Fusion Protein Ag12006 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |