Learn More
Epredia™ Frosted Microscope Slides
Premium slides manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to be used in routine histology and microbiology applications.
4.00€ - 4.40€
Specifications
Edges | 90°, Ground |
---|---|
Frosted | Yes |
Material | Soda Lime Glass |
Coating | Uncoated |
Writing Surface | Yes |
Description
- Manufactured from Swiss extra-white soda-lime glass
- Reduced iron content for low self-fluorescence
- Greenish shimmer minimized
- Guaranteed optimum, faultless transparency
- Clean, flat and suitable for immediate use
Specifications
90°, Ground | |
Soda Lime Glass | |
Yes |
Yes | |
Uncoated | |
76 x 26 mm |