Learn More
DOCK3/MOCA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20683-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Dedicator of cytokinesis3 (DOCK3), also named as MOCA and PBP, is a ∽180 kDa protein involved in signaling transduction. It is a potential guanine nucleotide exchange factor (GEF) which activate some small GTPases by exchanging bound GDP for free GTP. DOCK3 is associated in Alzheimer disease tangles and regulates the accumulation of amyloid precursor protein and beta-amyloid. Overexpression of Dock3 in neural cells promotes axonal outgrowth downstream of brain-derived neurotrophic factor (BDNF) signaling. DOCK3 binds to and inactivates glycogen synthase kinase-3beta (GSK-3beta) at the plasma membrane, thereby promoting axon branching and microtubule assembly. By stimulating actin polymerization and microtubule assembly, DOCK3 plays important roles downstream of BDNF signaling in the CNS.Spécification
DOCK3/MOCA | |
Polyclonal | |
Unconjugated | |
DOCK3 | |
dedicator of cytokinesis 3, DOCK3, KIAA0299, MOCA, Modifier of cell adhesion, PBP, Presenilin binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1795, 208869 | |
-20°C | |
Liquid |
Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CIQ7, Q8IZD9 | |
DOCK3 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |