Learn More
DDX43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17591-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.Spécification
DDX43 | |
Polyclonal | |
Unconjugated | |
DDX43 | |
Cancer/testis antigen 13, CT13, DDX43, DEAD box protein 43, DEAD box protein HAGE, HAGE, Helical antigen | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100048658, 55510, 682138 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NXZ2 | |
DDX43 | |
DDX43 Fusion Protein Ag11484 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |