Learn More
Agilent BioTek Cytation 1 Product Qualification Package - IQ/OQ/PQ
For Cytation 1 models
Brand:  Agilent BioTek 1650512
Product Code. 16522172
  • 832.00€ / Each
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Alert:
Note:
Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
Description
Description
  • Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
  • Comes on USB flash drive with the following contents:
    • Detailed required material list
    • Installation qualification procedure and checklist
    • Operational qualification procedure
    • Performance qualification procedure
    • Microsoft Excel spreadsheets for calculations required by procedures
    • Maintenance procedures
    • 5 year logbook for documenting all procedures/results
      • All of the documentation is available in both .doc and .pdf file format
Note:
  • Absorbance test plate (7260522) may be required in order to run some procedures specified for absorbance and multi-mode readers, but is not included and must be ordered separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytation 1 models
Detailed required material list, Installation qualification procedure and checklist, Operational qualification procedure, Performance qualification procedure, Microsoft Excel spreadsheets for calculations, Maintenance procedures, 5 year logbook