Learn More
Thermo Scientific™ Culti-Loops™ API™ NH QC Set
Description
Thermo Scientific™ Culti-Loops™ API NH Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Haemophilus influenzae Type b ATCC™ 10211™†
Haemophilus paraphrophilus ATCC™ 49917™†
Neisseria gonorrhoeae ATCC™ 31426™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
ATCC® | 10211TM* |
Description | API NH QC Set/3 |
Format | Culti-Loops |
Product Type | Culti-Loops QC Set |
Target Organism Class | Organism Sets |
Test Type | Qualitative QC |
Packaging Type | 1 Loop/Pack |
Quantity | 1/Pk. |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.