Learn More
Thermo Scientific™ Culti-Loops™ API™ 20 Strep QC Set
Product Code. 15289267
Click to view available options
Quantity:
5 Loops/Organism
Unit Size:
Each
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ API 20 Strep Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Streptococcus equi subsp. zooepidemicus group C ATCC™ 700400™†
Streptococcus uberisi ATCC™ 700407™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 700400TM*
Description API™ 20 Strep QC Set/2
Format Culti-Loops
Product Type Culti-Loops QC Set
Target Organism Class Organism Sets
Test Type Qualitative QC
Packaging Type 5 Loops/Package
Quantity 5 Loops/Organism
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.