Learn More
SNV Comines™ Katoenen laboratoriumjassen
Provides general light protection against splashes in the laboratory. SNV Comines™ Cotton Laboratory Coats are available in a range of sizes.
Merk: SNV Comines™ ANTLB00400T2
This item is not returnable.
View return policy
Beschrijving
- White in color
- Collared with lapels
- 4 buttons
Specificatie
Xavier-laboratoriumjas | |
Women's | |
Wit | |
4 | |
2 platte zakken, 1 borstzak, 1 binnenzak, openingshoek |
44 | |
100% katoen | |
Klik | |
96 cm | |
100% Cotton |