Learn More
BRE Mouse anti-Human, CoraLite 488, Clone: 2E5A5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL488-66356-100UL
Description
BRE, also named as BRCC45, is a death receptor-associated antiapoptotic protein, which is also involved in DNA-damage repair, and K63-specific deubiquitination. BRE and its XIAP-sustaining mechanism could represent novel targets for anti-cancer therapy. BRE may regulate TNF-alpha signaling through its interactions with TNFRSF1A. It plays a role in homeostasis or cellular differentiation in cells of neural, epithelial and germline origins. BRE gene has four isoforms with MW 42-47 kDa.
Spécification
BRE | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
IgG2a |
Immunofluorescence | |
2E5A5 | |
CoraLite 488 | |
100 μL | |
9577 | |
Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.