Learn More
Bel-Art™ Slide Boxes
Will not warp, splinter or crack, and are unaffected by moisture. Slide boxes are compact, durable slide boxes to provide the maximum protection for 25 x 75 mm (1 x 3 in.) microscope slides.
46.80€
Specifications
Type | Slide Box |
---|---|
For Use With (Equipment) | Microscope Slide |
Includes | Numbered index on base |
Product Code | Brand | Slide Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Slide Capacity | Price | Quantity & Availability | |||||
15515614
|
Bel-Art™
H44577-0000 |
100 | N/A | ||||||
Description
- Heavy-walled boxes are rugged and will not bend, split or chip
- Boxes are unaffected by humidity and moisture and are thoroughly insect-proof
- Easily identify slides with numbered index included on base and cover
Specifications
Slide Box | |
Microscope Slide | |
Numbered index on base |