Learn More
Deltalab™ Autoclave Resistant Bags
Wrap autoclaveable items in Deltalab™Autoclave Resistant Bags when working with laboratory items requiring sterilization.
44.04€ - 228.66€
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12004353
|
Deltalab™
200100 |
75 x 50cm |
44.04€
Each |
In Stock
Log in to see stock. ![]() |
|||||
12054363
|
Deltalab™
200310 |
30 x 60cm |
228.66€
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
12064363
|
Deltalab™
200311 |
40 x 75cm |
219.42€
Each |
In Stock
Log in to see stock. |
|||||
12074363
|
Deltalab™
200312 |
60 x 75cm |
205.00€
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
Description
- Printed with biohazard graphic symbol and use instructions in five languages
- ags placed in autoclave at 121ºC. should not be sealed hermetically
- Minimum order quantity: 50 pieces
Specifications
Autoclavable |