Learn More
anti-IgG2a, κ, PE-Cy5, Clone: G155-178, BD
Marque: BD Biosciences 555575
185.62 EUR valable jusqu'au 2025-12-31
Utilisez le code promo "25339" pour bénéficier de cette offre.
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry, Isotype Control
Spécification
Flow Cytometry | |
PE-Cyanine5 | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
G155-178 | |
Mouse | |
100 Tests | |
IgG2a κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.