Learn More
Sampling Systems™ Angular Beaker
These angular beaker attachments allow the cup to be set in specific locations while using the TeleScoop.
Marque: Sampling Systems™ 5065B-2002
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
- A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
- Attachments are interchangeable and simply click into position
Spécification
Swabbing Template | |
Autoclavable | |
Reusable | |
Non stérile |
Polypropylène | |
2 L | |
No |