Learn More
B Braun™ Aesculap™ Vega Cordless Clipper
An ideal product for veterinarians shaving small areas such as the back, neck, belly, head and face.
Brand: B Braun™ GT410
This item is not returnable.
View return policy
Description
- Very good cutting results thanks to clipping head made from a special steel alloy (carbon steel)
- Fits the hand perfectly thanks to slim shape
- Low weight of just 250 g including battery and clipping head
- Works very quietly and without vibration
- Can also be operated with charging cable connected
- 5 clip-on combs (4, 8, 12, 16, 20 mm)
- 3-way adjustable cutting lengths of 0.5, 1.5 and 2.5 mm
- Run time of 100 min, charging time of 60 min
Specifications
DC motor, approx. 4,800 strokes/min | |
Clippers | |
Carbon Steel | |
800 mAh, lithium-ion battery |
Veterinary Hair Cutting | |
Clipper, clipping head, 5 combs, 1 brush, 1 charger, 1 oil, 1 manual | |
Approx. 250 g |