missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADCY8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55065-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody is specific to ADCY8.
Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. The enzymatic activity is under the control of several hormones, and different polypeptides participate in the transduction of the signal from the receptor to the catalytic moiety. Stimulatory or inhibitory receptors (Rs and Ri) interact with G proteins (Gs and Gi) that exhibit GTPase activity and they modulate the activity of the catalytic subunit of the adenylyl cyclase.Spécification
ADCY8 | |
Polyclonal | |
Unconjugated | |
ADCY8 | |
AC8, ADCY3, ADCY8, adenylate cyclase 8 (brain), Adenylate cyclase type 8, Adenylate cyclase type VIII, Adenylyl cyclase 8, ATP pyrophosphate lyase 8, HBAC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114, 11514 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P40145, P97490 | |
ADCY8 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu