Learn More
Acetone, Basic, ACS, Scharlab
Product Code. 12426711
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE Bottle
Unit Size:
2.50 L
Brings a superb quality-price ratio to the laboratory. Acetone, Basic, ACS, Scharlab is a solvent that meets the specifications of the American Chemical Society.
The Basic product line is a selection of the most commonly used lab chemicals. These products are distinguished by their high quality, while at the same time ensuring the best prices.
Specifications
Melting Point -95°C
CAS 67-64-1
Color Colorless
Density 0.79g/cm3 at 20°C
Boiling Point 56°C
Quantity 2.5 L
Flash Point −20°
Ignition Point 540°C
Packaging HDPE Bottle
Physical Form Liquid
Refractive Index 1.3588
Molecular Formula C3H6O
Linear Formula (CH3)2CO
MDL Number MFCD00008765
Vapor Pressure 233 hPa at 20°C
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Solubility Information Miscible in water
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Grade Basic
Chemical Name or Material Acetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5LT Acetone, ACS BASIC
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetone