Learn More
Agilent BioTek 8 Channel Manifolds
8-Channel Manifold, for 96-well Half Area and PCR Microplates
Brand: Agilent BioTek 7140515S
This item is not returnable.
View return policy
Description
To be used with MicroFill (AF1000A)
Specifications
8-channel Manifold Dispensing Comb | |
Autoclavable | |
96 Well Half Area and PCR Microplates |