Learn More
Agilent BioTek 8 Channel Manifolds
8-Channel Manifold, for 24-well Microplates
Brand: Agilent BioTek 7140518S
This item is not returnable.
View return policy
Description
To be used with MicroFill (AF1000A)
Specifications
8-channel Manifold Dispensing Comb | |
Autoclavable | |
24 Well Microplates |