missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSTR5 Mouse anti-Human, Mouse, Rat, Clone: 2B10D8, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Spécification
Antigène | SSTR5 |
---|---|
Clone | 2B10D8 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16868955
|
Proteintech
66772-1-IG-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16858955
|
Proteintech
66772-1-IG-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predomitly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.Spécification
SSTR5 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
somatostatin receptor 5, Somatostatin receptor type 5, SS 5 R, SS5 R, SS5R, SSTR5 | |
SSTR5 | |
IgG1 | |
Protein G | |
Antibody |
2B10D8 | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat, Mouse, Human | |
O08858, P30938, P35346 | |
20609, 25354, 6755 | |
SSTR5 Fusion Protein Ag18615 | |
Primary | |
-20°C | |
SSTR5 |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
SSTR5 Mouse anti-Human, Mouse, Rat, Clone: 2B10D8, Proteintech