Learn More
GDI1 Mouse anti-Human, CoraLite 594, Clone: 4E1G1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL594-66434-100UL
Description
The GDI1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GDI1. This antibody recognizes human, mouse, rat, pig antigen. The GDI1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spécification
GDI1 | |
Monoclonal | |
CoraLite 594 | |
100 μL | |
2664 | |
IgG1 |
Immunofluorescence | |
4E1G1 | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.