Learn More
CoraLite™ Plus 488-conjugated S100A6 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech CL488-10245-20UL
Code produit. 30235796
  • 129.84€ / 20µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
The S100A6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human S100A6. This antibody recognizes human antigen. The S100A6 antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.
Spécification
Spécification
S100A6
Polyclonal
CoraLite Plus 488
Rabbit
Antigen affinity purification
RUO
6277
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining)
1000 μg/mL
2A9; 5B10; CABP; CACY; PRA
Recombinant Protein
20 μL
Primary
Human
Conjugated Antibody