Learn More
CoraLite™ Plus 405 Anti-Human CD11c (3.9), Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech CL405-65086-100TESTS
Code produit. 30237459
  • 128.83€ / 1 kit
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
The CD11c antibody from Proteintech is a mouse monoclonal antibody generated with rheumatoid synovial cells and human monocytes of human CD11c. This antibody recognizes human antigen. The CD11c antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 405.
Spécification
Spécification
CD11c
Monoclonal
5 μl/test
CD11C; SLEB6
Rheumatoid synovial cells and human monocytes
100 Tests
Primary
Human
Flow Antibody
IgG1
Flow Cytometry
3.9
CoraLite Plus 405
Mouse
Affinity purification
RUO
3687
Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment.