Learn More
CALD1 Mouse anti-Human, CoraLite 594, Clone: 1E1C11, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL594-66693-100UL
Description
The CALD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CALD1. This antibody recognizes human, mouse, rat antigen. The CALD1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Spécification
CALD1 | |
Monoclonal | |
CoraLite 594 | |
100 μL | |
800 | |
IgG2b |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
1E1C11 | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.